CAS RN 81-15-2
IUPAC Navn:
1-tert-butyl-3,5-dimethyl-2,4,6-trinitrobenzene
Synonymer:
musk xylene
Molekylvægt (g/mol):
297.27
Molekylformel:
C12H15N3O6
InChi Key:
XMWRWTSZNLOZFN-UHFFFAOYSA-N
SMILES:
CC1=C(C(=C(C(C)=C1[N+]([O-])=O)[N+]([O-])=O)C(C)(C)C)[N+]([O-])=O
1
–
3
af
3
Resultater
1
Musk xylene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Musk xylene 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
Musk xylene 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality