CAS RN 80981-00-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(S)-8-bis[(Phenylmethoxy)carbonyl]-3-imino-12,12-dimethyl-10-oxo-4-phenylmethyl Ester 11-Oxa-2,4,9-triazatridecanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-8-bis[(Phenylmethoxy)carbonyl]-3-imino-12,12-dimethyl-10-oxo-4-phenylmethyl Ester 11-Oxa-2,4,9-triazatridecanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-8-bis[(Phenylmethoxy)carbonyl]-3-imino-12,12-dimethyl-10-oxo-4-phenylmethyl Ester 11-Oxa-2,4,9-triazatridecanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.