CAS RN 802904-66-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
AZD1981, MedChemExpress
MedChemExpress AZD1981 is a potent and selective CRTh2 antagonist; displaces radio-labelled PGD2 from human recombinant DP2 with high potency (pIC50 = 8.4).