CAS RN 787618-22-8
IUPAC Navn:
[2',6'-bis(propan-2-yloxy)-[1,1'-biphenyl]-2-yl]dicyclohexylphosphane
Synonymer:
dicyclohexyl({2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl})phosphane
Molekylvægt (g/mol):
466.65
Molekylformel:
C30H43O2P
InChi Key:
MXFYYFVVIIWKFE-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=CC(OC(C)C)=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
1
–
3
af
3
Resultater
1
2-(Dicyclohexylphosphino)-2',6'-di-isopropoxy-1,1'-biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Dicyclohexylphosphino)-2',6'-di-isopropoxy-1,1'-biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Dicyclohexylphosphino)-2',6'-di-isopropoxy-1,1'-biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.