CAS RN 78761-59-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
11Alpha,17Alpha,21-Trihydroxy-16Alpha-methyl-1,4-pregnadiene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
11Alpha,17Alpha,21-Trihydroxy-16Alpha-methyl-1,4-pregnadiene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
11Alpha,17Alpha,21-Trihydroxy-16Alpha-methyl-1,4-pregnadiene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.