CAS RN 78715-23-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(2S)-2-(4,5alpha-Epoxy-3-hydroxy-6-methoxy-6alpha,14-ethano-14alpha-morphinan-7alpha-yl)-3,3-dimethylbutan-2-ol (Norbuprenorphine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.