CAS RN 78-73-9
IUPAC Navn:
(2-hydroxyethyl)trimethylazanium hydrogen carbonate
Synonymer:
choline bicarbonate
Molekylvægt (g/mol):
165.19
Molekylformel:
C6H15NO4
InChi Key:
DQKGOGJIOHUEGK-UHFFFAOYSA-M
SMILES:
OC([O-])=O.C[N+](C)(C)CCO
1
–
3
af
3
Resultater
1
Choline Bicarbonate (80% in water), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Choline Bicarbonate (80% in water), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Choline Bicarbonate (80% in water), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.