CAS RN 76-06-2
IUPAC Navn:
trichloro(nitro)methane
Synonymer:
chloropicrin trichloronitromethane (chloropicrine) chlorpikrin 1,1,1-trichloronitromethane
Molekylvægt (g/mol):
164.37
Molekylformel:
CCl3NO2
InChi Key:
LFHISGNCFUNFFM-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C(Cl)(Cl)Cl
1
–
2
af
2
Resultater
1
Chloropicrin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality