CAS RN 757251-39-1
IUPAC Navn:
4-(4-amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide
Synonymer:
4-(4-amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide
Molekylvægt (g/mol):
261.26
Molekylformel:
C13H12FN3O2
InChi Key:
ZQHJPIPGBODVTG-UHFFFAOYSA-N
SMILES:
CNC(=O)C1=NC=CC(OC2=CC(F)=C(N)C=C2)=C1
1
–
4
af
4
Resultater
1
4-(4-Amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(4-Amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(4-Amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
4-(4-Amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.