CAS RN 752-56-7
IUPAC Navn:
(2R,3S,4S)-1,2,4-tris(butanoyloxy)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}pentan-3-yl butanoate
Synonymer:
riboflavin tetrabutyrate
Molekylvægt (g/mol):
656.73
Molekylformel:
C33H44N4O10
InChi Key:
MJNIWUJSIGSWKK-BBANNHEPSA-N
SMILES:
CCCC(=O)OC[C@@H](OC(=O)CCC)[C@@H](OC(=O)CCC)[C@H](CN1C2=CC(C)=C(C)C=C2N=C2C(=O)NC(=O)N=C12)OC(=O)CCC
1
–
4
af
4
Resultater
1
Riboflavine Tetrabutyrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Riboflavine Tetrabutyrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Riboflavine Tetrabutyrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Riboflavin Tetrabutyrate, MedChemExpress
MedChemExpress Riboflavin Tetrabutyrate is a lipophilic flavin derivative with antioxidative and lipid peroxide-removing activity.