CAS RN 7517-36-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Monocyclohexyl Phthalate, TRC
CAS: 7517-36-4 Molekylær formel: C14 H16 O4 Molekylvægt (g/mol): 248.27 Synonym: 1,2-Benzenedicarboxylic acid, monocyclohexyl ester (9CI),Phthalic acid, cyclohexyl ester (6CI,7CI),Phthalic acid, monocyclohexyl ester (8CI),Cyclohexyl hydrogen phthalate,Monocyclohexyl phthalate IUPAC navn: 2-cyclohexyloxycarbonylbenzoic acid SMIL: OC(=O)c1ccccc1C(=O)OC2CCCCC2