CAS RN 74389-68-7
IUPAC Navn:
(2-{[(2R)-2-(acetyloxy)-3-(hexadecyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonymer:
platelet activating factor platelet-activating factor C16-paf C16-paf acether
Molekylvægt (g/mol):
523.69
Molekylformel:
C26H54NO7P
InChi Key:
HVAUUPRFYPCOCA-AREMUKBSSA-N
SMILES:
CCCCCCCCCCCCCCCCOC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(C)=O