CAS RN 73960-72-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Acetyl-4,7,8,9-tetra-O-acetyl-2,3-dehydro-2-deoxyneuraminic Acid, Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Acetyl-4,7,8,9-tetra-O-acetyl-2,3-dehydro-2-deoxyneuraminic Acid, Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Acetyl-4,7,8,9-tetra-O-acetyl-2,3-dehydro-2-deoxyneuraminic Acid, Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.