CAS RN 73851-70-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Ranitidine S-Oxide, TRC
CAS: 73851-70-4 Molekylær formel: C13 H22 N4 O4 S Molekylvægt (g/mol): 330.4 Synonym: N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine,Ranitidine Hydrochloride Imp. C (EP),Ranitidine Sulphoxide IUPAC navn: 1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine SMIL: CNC(=C[N+](=O)[O-])NCCS(=O)Cc1oc(CN(C)C)cc1
2
N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine (Ranitidine Sulphoxide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.