CAS RN 73215-11-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1-(1-Ethylpropyl)-5,6-dimethyl-7-nitro-1H-benzimidazole, TRC
CAS: 73215-11-9 Molekylær formel: C14 H19 N3 O2 Molekylvægt (g/mol): 261.3196 Synonym: 1H-Benzimidazole, 1-(1-ethylpropyl)-5,6-dimethyl-7-nitro-,1-(1-Ethylpropyl)-5,6-dimethyl-7-nitro-1H-benzimidazole IUPAC navn: 5,6-dimethyl-7-nitro-1-pentan-3-ylbenzimidazole SMIL: CCC(CC)n1cnc2cc(C)c(C)c(c12)[N+](=O)[O-]
2
Pendimethalin metabolite 1 M455H029 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality