CAS RN 729-46-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ JX 06
Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor
2
3