CAS RN 720675-74-1
IUPAC Navn:
2-(2,4-dimethoxyphenyl)-6-methoxy-4H-chromen-4-one
Synonymer:
2-(2,4-dimethoxyphenyl)-6-methoxychromen-4-one
Molekylvægt (g/mol):
312.32
Molekylformel:
C18H16O5
InChi Key:
WUWFDVDASNSUKP-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(C=C1)C1=CC(=O)C2=CC(OC)=CC=C2O1
1
–
3
af
3
Resultater
1
6,2',4'-Trimethoxyflavone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6,2',4'-Trimethoxyflavone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6,2',4'-Trimethoxyflavone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.