CAS RN 7206-76-0
IUPAC Navn:
2-ethyl-2-phenylpropanediamide
Synonymer:
2 ethyl 2 phenylmalonamide
Molekylvægt (g/mol):
206.25
Molekylformel:
C11H14N2O2
InChi Key:
JFZHPFOXAAIUMB-UHFFFAOYSA-N
SMILES:
CCC(C(N)=O)(C(N)=O)C1=CC=CC=C1
1
–
3
af
3
Resultater
1
2-Ethyl-2-phenylmalonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Ethyl-2-phenylmalonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Ethyl-2-phenylmalonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.