CAS RN 71208-12-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Phenylmethyl 2-(Acetylamino)-2-deoxy-6-O-(phenylmethyl)-3-O-(2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phenylmethyl 2-(Acetylamino)-2-deoxy-6-O-(phenylmethyl)-3-O-(2,3,4,6-tetra-O-acetyl-b-D-galactopyranosyl)-a-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Phenylmethyl 2-(Acetylamino)-2-deoxy-6-O-(phenylmethyl)-3-O-(2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.