Få mere at vide
(±)-Mandelic-2,3,4,5,6-d5 Acid, CDN
Artikelnummer. 30328654
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328654 0.5 g 0.5g
30325496 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328654 Leverandør CDN Leverandørnr. CDND63540.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Mandelic-2,3,4,5,6-d5 Acid
CAS 70838-71-0
Molekylær formel C8 2H5 H3 O3
Mængde 0.5 g
InChI formel InChI=1 S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9 H,(H,10,11)/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(O)C(=O)O)c([2 H])c1[2 H]
IUPAC navn 2-hydroxy-2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid
Molekylvægt (g/mol) 157.18
Formel vægt 157.0787 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity