CAS RN 70831-56-0
IUPAC Navn:
(2R,3R)-2,3-bis({[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy})butanedioic acid
Synonymer:
cichoric acid
Molekylvægt (g/mol):
474.37
Molekylformel:
C22H18O12
InChi Key:
YDDGKXBLOXEEMN-IABMMNSOSA-N
SMILES:
OC(=O)[C@H](OC(=O)\C=C\C1=CC=C(O)C(O)=C1)[C@@H](OC(=O)\C=C\C1=CC=C(O)C(O)=C1)C(O)=O
1
–
1
af
1
Resultater
1
L-Chicoric Acid, MedChemExpress
MedChemExpress L-Chicoric Acid ((-)-Chicoric acid) is a dicaffeoyltartaric acid and a potent, selective and reversible HIV-1 integrase inhibitor with an IC50 of ∼100 nM. L-Chicoric Acid inhibits HIV-1 replication in tissue culture.