CAS RN 707-99-3
IUPAC Navn:
2-(6-amino-9H-purin-9-yl)ethan-1-ol
Synonymer:
2-(6-aminopurin-9-yl)ethanol
Molekylvægt (g/mol):
179.18
Molekylformel:
C7H9N5O
InChi Key:
VAQOTZQDXZDBJK-UHFFFAOYSA-N
SMILES:
NC1=C2N=CN(CCO)C2=NC=N1
1
–
3
af
3
Resultater
1
2-(6-Aminopurin-9-yl)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(6-Aminopurin-9-yl)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(6-Aminopurin-9-yl)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.