CAS RN 706782-28-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Pimavanserin hemitartrate, MedChemExpress
MedChemExpress Pimavanserin (ACP-103) hemitartrate is a potent 5-HT 2A receptor inverse agonist with pIC50 and pKi of 8.73 and 9.3, respectively.