CAS RN 7059-24-7
IUPAC Navn:
(2R,3S,4R,6S)-6-{[(6S,7S)-6-{[(2S,4R,5R,6R)-4-{[(2S,4R,5R,6R)-4-{[(2S,4S,5S,6S)-5-(acetyloxy)-4-hydroxy-4,6-dimethyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-7-[(1S,3S,4R)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-4,10-dihydroxy-3-methyl-5-oxo-5,6,7,8-tetrahydroanthracen-2-yl]oxy}-4-{[(2R,4R,5R,6R)-4-hydroxy-5-methoxy-6-methyloxan-2-yl]oxy}-2-methyloxan-3-yl acetate
Synonymer:
chromomycin A3 chromomycin A(sub 3) antibiotic 69895 A toyomicin toyomycin chromomycin a-3 aburamycin B antibiotic from streptomyces griseus antibiotic b 599 antibiotic B 599
Molekylvægt (g/mol):
1183.26
Molekylformel:
C57H82O26
InChi Key:
ZYVSOIYQKUDENJ-WKSBCEQHSA-N
SMILES:
CO[C@@H]([C@@H]1CC2=CC3=CC(O[C@H]4C[C@@H](O[C@@H]5C[C@@H](O)[C@@H](OC)[C@@H](C)O5)[C@@H](OC(C)=O)[C@@H](C)O4)=C(C)C(O)=C3C(O)=C2C(=O)[C@H]1O[C@H]1C[C@@H](O[C@H]2C[C@@H](O[C@H]3C[C@](C)(O)[C@@H](OC(C)=O)[C@H](C)O3)[C@H](O)[C@@H](C)O2)[C@H](O)[C@@H](C)O1)C(=O)[C@@H](O)[C@@H](C)O