CAS RN 701-54-2
IUPAC Navn:
4-(aminomethyl)cyclohexane-1-carboxylic acid
Synonymer:
tranexamic acid
Molekylvægt (g/mol):
157.21
Molekylformel:
C8H15NO2
InChi Key:
GYDJEQRTZSCIOI-UHFFFAOYSA-N
SMILES:
NCC1CCC(CC1)C(O)=O
1
–
3
af
3
Resultater
1
4-(Aminomethyl)cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Aminomethyl)cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(Aminomethyl)cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.