CAS RN 69999-16-2
IUPAC Navn:
2-(2,3-dihydro-1-benzofuran-5-yl)acetic acid
Synonymer:
2,3-dihydro-1-benzofuran-5-ylacetic acid
Molekylvægt (g/mol):
178.19
Molekylformel:
C10H10O3
InChi Key:
LALSYIKKTXUSLG-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC2=C(OCC2)C=C1
1
–
3
af
3
Resultater
1
2,3-Dihydro-5-benzofuranacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3-Dihydro-5-benzofuranacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3-Dihydro-5-benzofuranacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.