CAS RN 695207-56-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ BC-LI-0186
Leucyl-tRNA synthase (TRS)/Ras-related GTP-binding protein D (RagD) interaction inhibitor