CAS RN 6938-94-9
IUPAC Navn:
1,6-bis(propan-2-yl) hexanedioate
Synonymer:
diisopropyl adipate wickenol 116 crodamol DA standamul dipa prodipate tegester 504-D β dia schercemol dia ceraphyl 230 bis(1-methylethyl)hexanedioate
Molekylvægt (g/mol):
230.3
Molekylformel:
C12H22O4
InChi Key:
ZDQWESQEGGJUCH-UHFFFAOYSA-N
SMILES:
CC(C)OC(=O)CCCCC(=O)OC(C)C
1
–
1
af
1
Resultater
1
Diisopropyl Adipate, TRC
CAS: 6938-94-9 Molekylær formel: C12 H22 O4 Molekylvægt (g/mol): 230.3 Synonym: Ceraphyl 230,Crodamol DA,Diisopropyl Adipate,Iso Adipat,Isopropyl Adipate,NSC 56587,Nikkol DID,Schercemol DIA,Standamul DIPA,Wickenol 116; IUPAC navn: diisopropyl hexanedioate SMIL: CC(C)OC(=O)CCCCC(=O)OC(C)C