CAS RN 69327-76-0
IUPAC Navn:
2-(tert-butylimino)-5-phenyl-3-(propan-2-yl)-1,3,5-thiadiazinan-4-one
Synonymer:
buprofezin
Molekylvægt (g/mol):
305.44
Molekylformel:
C16H23N3OS
InChi Key:
PRLVTUNWOQKEAI-UHFFFAOYSA-N
SMILES:
CC(C)N1C(SCN(C1=O)C1=CC=CC=C1)=NC(C)(C)C
1
–
4
af
4
Resultater
1
Buprofezin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Buprofezin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
GB 23200.121-2021 Pesticide Mixture 10 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality