CAS RN 690211-40-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(1R,6R)-Benzyl 2-(1,2-Dihydroxypropan-2-yl)-9-azabicyclo[4.2.1]non-2-ene-9-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1R,6R)-Benzyl 2-(1,2-Dihydroxypropan-2-yl)-9-azabicyclo[4.2.1]non-2-ene-9-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1R,6R)-Benzyl 2-(1,2-Dihydroxypropan-2-yl)-9-azabicyclo[4.2.1]non-2-ene-9-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.