CAS RN 6823-83-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Chloroquine Sulfate Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Chloroquine Sulfate, TRC
CAS: 6823-83-2 Molekylær formel: C18H26ClN3.H2O.H2O4S Molekylvægt (g/mol): 435.97 Synonym: Chloroquine sulfate,N4-(7-chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine sulfate IUPAC navn: 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;sulfuric acid;hydrate SMIL: O.CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OS(=O)(=O)O