CAS RN 67684-64-4
IUPAC Navn:
(1R,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Synonymer:
(1R,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Molekylvægt (g/mol):
173.17
Molekylformel:
C7H11NO4
InChi Key:
YFYNOWXBIBKGHB-CLZZGJSISA-N
SMILES:
N[C@@]1(CC[C@H](C1)C(O)=O)C(O)=O
1
–
2
af
2
Resultater
1
Tocris Bioscience™ (±)-trans-ACPD
CAS: 67684-64-4 Molekylær formel: C7H11NO4
2
trans-ACPD, MedChemExpress
MedChemExpress trans-ACPD, a metabotropic receptor agonist, produces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons.