CAS RN 66761-27-1
IUPAC Navn:
3-azido-5-hydroxybenzoic acid
Synonymer:
3-azido-5-hydroxybenzoic acid
Molekylvægt (g/mol):
179.14
Molekylformel:
C7H5N3O3
InChi Key:
IMKRSVNNWZYMKI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(O)=CC(=C1)N=[N+]=[N-]
1
–
3
af
3
Resultater
1
4-Azidosalicylic Acid(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Azidosalicylic Acid(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Azidosalicylic Acid(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.