CAS RN 666844-61-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3N-Benzyloxycarbonyl 3-Amino-2,2-dimethylpropanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3N-Benzyloxycarbonyl 3-Amino-2,2-dimethylpropanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3N-Benzyloxycarbonyl 3-Amino-2,2-dimethylpropanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.