CAS RN 654671-78-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Sitagliptin Phosphate, TRC
CAS: 654671-78-0 Molekylær formel: C16 H15 F6 N5 O . H3 O4 P Molekylvægt (g/mol): 505.31 IUPAC navn: (3R)-3-amino-1-[3-(trifluoromethyl)-6,8-dihydro-5H-[1,2,4]triazolo[4,3-a]pyrazin-7-yl]-4-(2,4,5-trifluorophenyl)butan-1-one;phosphoric acid SMIL: N[C@@H](CC(=O)N1CCn2c(C1)nnc2C(F)(F)F)Cc3cc(F)c(F)cc3F.OP(=O)(O)O
2
Tocris Bioscience™ Sitagliptin Phosphate
Potent and selective DPP IV (DPP4) inhibitor (IC50 = 18 nM)
3
Sitagliptin phosphate, MedChemExpress
MedChemExpress Sitagliptin phosphate (MK-0431 phosphate) is a potent inhibitor of DPP4 with an IC50 of 19 nM in Caco-2 cell extracts.