CAS RN 65371-16-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
6-Azido-6-deoxy-1,2-O-isopropylidene-Alpha-D-glucofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Azido-6-deoxy-1,2-O-isopropylidene-Alpha-D-glucofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Azido-6-deoxy-1,2-O-isopropylidene-Alpha-D-glucofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.