CAS RN 64603-90-3
IUPAC Navn:
hydrogen 1,2,3,6-tetrahydropyridine-4-carboxylic acid chloride
Synonymer:
isoguvacine
Molekylvægt (g/mol):
163.6
Molekylformel:
C6H10ClNO2
InChi Key:
SUWREQRNTXCCBL-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].OC(=O)C1=CCNCC1
1
–
2
af
2
Resultater
1
1,2,3,6-Tetrahydropyridine-4-carboxylic Acid (Isoguvacine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.