CAS RN 6427-66-3
IUPAC Navn:
4-azidobenzoic acid
Synonymer:
4-azidobenzoic acid
Molekylvægt (g/mol):
163.14
Molekylformel:
C7H5N3O2
InChi Key:
PQXPAFTXDVNANI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C=C1)N=[N+]=[N-]
1
–
3
af
3
Resultater
1
4-Azidobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Azidobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Azidobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.