CAS RN 636-95-3
IUPAC Navn:
hydrogen (3-nitrophenyl)hydrazine chloride
Synonymer:
hydrogen (3-nitrophenyl)hydrazine chloride
Molekylvægt (g/mol):
189.6
Molekylformel:
C6H8ClN3O2
InChi Key:
BKOYKMLGFFASBG-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NNC1=CC=CC(=C1)[N+]([O-])=O
1
–
2
af
2
Resultater
1
3-Nitrophenylhydrazine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Nitrophenylhydrazine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.