CAS RN 6293-83-0
IUPAC Navn:
2-iodo-4-nitroaniline
Synonymer:
2-iodo-4-nitroaniline
Molekylvægt (g/mol):
264.02
Molekylformel:
C6H5IN2O2
InChi Key:
LOLSEMNGXKAZBZ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1I)[N+]([O-])=O
1
–
3
af
3
Resultater
1
2-Iodo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Iodo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Iodo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.