CAS RN 62873-16-9
IUPAC Navn:
5-methyl-3-methylideneoxolan-2-one
Synonymer:
5-methyl-3-methylideneoxolan-2-one
Molekylvægt (g/mol):
112.13
Molekylformel:
C6H8O2
InChi Key:
KYLUHLJIAMFYKW-UHFFFAOYNA-N
SMILES:
CC1CC(=C)C(=O)O1
1
–
1
af
1
Resultater
1
alpha-Methylene-gamma-valerolactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.