CAS RN 6271-79-0
IUPAC Navn:
3,3'-dinitro-[1,1'-biphenyl]-4,4'-diamine
Synonymer:
3,3'-dinitro-[1,1'-biphenyl]-4,4'-diamine
Molekylvægt (g/mol):
274.24
Molekylformel:
C12H10N4O4
InChi Key:
OCEINMLGYDSKFW-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1[N+]([O-])=O)C1=CC=C(N)C(=C1)[N+]([O-])=O
1
–
2
af
2
Resultater
1
3,3’-Dinitrobenzidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,3’-Dinitrobenzidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.