CAS RN 627-59-8
IUPAC Navn:
5-methylhexan-2-ol
Synonymer:
5-methyl-2-hexanol
Molekylvægt (g/mol):
116.2
Molekylformel:
C7H16O
InChi Key:
ZDVJGWXFXGJSIU-UHFFFAOYNA-N
SMILES:
CC(C)CCC(C)O
1
–
3
af
3
Resultater
1
5-Methyl-2-hexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Methyl-2-hexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Methyl-2-hexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.