CAS RN 626-89-1
IUPAC Navn:
4-methylpentan-1-ol
Synonymer:
4-methyl-1-pentanol
Molekylvægt (g/mol):
102.18
Molekylformel:
C6H14O
InChi Key:
PCWGTDULNUVNBN-UHFFFAOYSA-N
SMILES:
CC(C)CCCO
1
–
3
af
3
Resultater
1
Isohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Isohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Isohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.