CAS RN 6236-09-5
IUPAC Navn:
(2S)-2-hydroxy-2-methylbutanedioic acid
Synonymer:
pyrotartaric acid (S)-citramalate S-citramalic acid (S)-citramalic acid L-citramalic acid MEZ pyrotartarate (3S)-citramalic acid L-α-hydroxypyrotartaric acid S-α-hydroxypyrotartaric acid
Molekylvægt (g/mol):
148.11
Molekylformel:
C5H8O5
InChi Key:
XFTRTWQBIOMVPK-YFKPBYRVSA-N
SMILES:
C[C@](O)(CC(O)=O)C(O)=O
1
–
3
af
3
Resultater
1
(+)-Citramalic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-Citramalic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+)-Citramalic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.