CAS RN 620-12-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Glyceryl Mononitrate Solution [1.0mg/mL in Acetonitrile], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Glyceryl Mononitrate Solution [100μg/mL, in Acetonitrile], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.