CAS RN 615-66-7
IUPAC Navn:
2-chlorobenzene-1,4-diamine; sulfuric acid
Synonymer:
2-chloro-P-phenylenediamine
Molekylvægt (g/mol):
240.66
Molekylformel:
C6H9ClN2O4S
InChi Key:
GQFGHCRXPLROOF-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1
1
–
3
af
3
Resultater
1
2-Chlorobenzene-1,4-diamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Chlorobenzene-1,4-diamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chlorobenzene-1,4-diamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.