CAS RN 61337-68-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine (Desmethylmirtazapine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.