CAS RN 61301-33-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Schisandrin C, MedChemExpress
MedChemExpress Schisandrin C (Schizandrin-C) is a phytochemical lignan isolated from Schizandra chinensis. Schisandrin C has diverse biological activities, including anticancer, anti-inflammatory and antioxidant effects. Schisandrin C can be used for cancer, alzheimer’s disease, and liver diseases research. Schisandrin C induces cell apoptosis.