CAS RN 6124-79-4
IUPAC Navn:
4-methyl-2,5-dihydrofuran-2-one
Synonymer:
4-methyl-5H-furan-2-one
Molekylvægt (g/mol):
98.1
Molekylformel:
C5H6O2
InChi Key:
ZZEYQBNQZKUWKY-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)OC1
1
–
3
af
3
Resultater
1
4-Methyl-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methyl-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methyl-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.